Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Flot1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022701.2 | NP_073192.2 | 3, 4 | 300 | ||
XM_006256024.2 | XP_006256086.1 | 3, 4 | 319 | |||
GenBank mRNA | BC064652.1 | - | 3, 4 | 299 | 5598180 | |
CK359020.1 | - | 3, 4 | 300 | |||
U60976.1 | - | 3, 4 | 230 | |||
U60977.1 | - | 2, 3 | 304 |