Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sfxn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022948.1 | NP_075237.1 | 3 | 495 | ||
XM_006231563.2 | XP_006231625.1 | 4 | 645 | |||
XM_006231565.2 | XP_006231627.1 | 3 | 363 | |||
XM_006231566.2 | XP_006231628.1 | 3 | 385 | |||
XM_008760451.1 | XP_008758673.1 | 3 | 382 | |||
GenBank mRNA | AF276997.1 | - | 3 | 495 | ||
BC062043.1 | - | 3 | 524 | 5597953 |