Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC306079 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047879.1 | NP_001041344.1 | 2 | 575 | ||
GenBank mRNA | AY387052.1 | - | 2 | 575 |
Gene Symbol: Rpl14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022949.2 | NP_075238.2 | 3 | 174 | ||
GenBank mRNA | BC168728.1 | - | 3 | 174 | 6918710 | |
X94242.1 | - | 2 | 171 |