Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022699.3 | NP_073190.1 | 4, 5 | 379 | ||
GenBank mRNA | BC058471.1 | - | 4, 5 | 383 | 6919868 | |
FQ210232.1 | - | 4, 5 | 376 | |||
FQ210454.1 | - | 4, 5 | 375 | |||
FQ210629.1 | - | 4, 5 | 379 | |||
FQ211005.1 | - | 4, 5 | 381 | |||
FQ211389.1 | - | 4, 5 | 377 | |||
FQ211694.1 | - | 4, 5 | 376 | |||
FQ211731.1 | - | 4, 5 | 375 | |||
FQ211953.1 | - | 4, 5 | 379 | |||
FQ214466.1 | - | 4, 5 | 376 | |||
FQ217830.1 | - | 4, 5 | 376 | |||
FQ217913.1 | - | 4, 5 | 376 | |||
FQ221046.1 | - | 4, 5 | 376 | |||
FQ221166.1 | - | 4, 5 | 376 | |||
FQ221653.1 | - | 4, 5 | 384 | |||
FQ221662.1 | - | 4, 5 | 377 | |||
FQ221710.1 | - | 4, 5 | 376 | |||
FQ221740.1 | - | 4, 5 | 375 | |||
FQ221836.1 | - | 4, 5 | 376 | |||
FQ222551.1 | - | 4, 5 | 376 | |||
FQ222688.1 | - | 4, 5 | 375 | |||
FQ222703.1 | - | 4, 5 | 375 | |||
FQ223191.1 | - | 4, 5 | 375 | |||
FQ223319.1 | - | 4, 5 | 376 | |||
FQ223321.1 | - | 4, 5 | 357 | |||
FQ223373.1 | - | 4, 5 | 375 | |||
FQ223410.1 | - | 4, 5 | 473 | |||
FQ223428.1 | - | 4, 5 | 323 | |||
FQ223604.1 | - | 696 | ||||
FQ223661.1 | - | 4, 5 | 376 | |||
FQ224433.1 | - | 4, 5 | 378 | |||
FQ224506.1 | - | 4, 5 | 375 | |||
FQ224779.1 | - | 4, 5 | 376 | |||
FQ228655.1 | - | 4, 5 | 375 | |||
FQ228991.1 | - | 4, 5 | 375 | |||
FQ229525.1 | - | 4, 5 | 374 | |||
FQ229603.1 | - | 976 | ||||
K02932.1 | - | 3, 4 | 306 |
Gene Symbol: LOC100362027 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002727709.3 | XP_002727755.1 | 1 | 383 |