Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022700.2 | NP_073191.1 | 2 | 231 | ||
XM_006231559.2 | XP_006231621.1 | 3 | 752 | |||
GenBank mRNA | BC084722.1 | - | 2 | 231 | 7310623 | |
U12568.1 | - | 2 | 235 | |||
X76921.1 | - | 3 | 363 |