Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004224.3 | NP_004215.2 | 2 | 1508 | ||
XM_011531216.2 | XP_011529518.1 | 2 | 737 | |||
GenBank mRNA | BC103696.1 | - | 2 | 1418 | 40006713 | |
BC105683.1 | - | 2 | 1418 | 40006718 | ||
BC105684.1 | - | 2 | 1418 | 40006715 | ||
EU432118.1 | - | 2 | 1370 | |||
U52219.1 | - | 2 | 1418 |
Set Membership: |
G-protein coupled receptor |