Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olah | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022705.1 | NP_073196.1 | 5 | 669 | ||
XM_008771886.1 | XP_008770108.1 | 6 | 823 | |||
XM_008771887.1 | XP_008770109.1 | 5 | 526 | |||
GenBank mRNA | M16200.1 | - | 5 | 669 | ||
Y00311.1 | - | 4 | 404 |