Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mvp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022715.2 | NP_073206.2 | 2, 3 | 183 | ||
GenBank mRNA | BC071174.1 | - | 2, 3 | 190 | 7099451 | |
U09870.2 | - | 2, 3 | 183 |