Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLP2R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004246.2 | NP_004237.1 | 2, 3 | 788 | ||
XM_005256861.3 | XP_005256918.1 | 1, 2 | 86 | |||
XM_011524077.2 | XP_011522379.1 | 2, 3 | 827 | |||
XM_017025339.1 | XP_016880828.1 | 2, 3 | 827 | |||
GenBank mRNA | AF105367.1 | - | 2, 3 | 275 | ||
AK295765.1 | - | 2, 3 | 167 | |||
AK314269.1 | - | 2, 3 | 788 | |||
BC096261.1 | - | 2, 3 | 275 | |||
BC096262.1 | - | 2, 3 | 275 | 40006110 | ||
BC096263.1 | - | 2, 3 | 271 | |||
BC096264.1 | - | 2, 3 | 275 |
Set Membership: |
G-protein coupled receptor |