Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LY86 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004271.3 | NP_004262.1 | 3, 4 | 386 | ||
GenBank mRNA | AB020499.1 | - | 2, 3 | 222 | ||
AF057178.2 | - | 3, 4 | 375 | |||
BC038846.1 | - | 3, 4 | 396 | 6015123 | ||
CD695164.1 | - | 3, 4 | 450 |