Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BAG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004281.3 | NP_004272.2 | 4 | 2170 | ||
XM_005270287.1 | XP_005270344.1 | 4 | 2190 | |||
GenBank mRNA | AF071218.2 | - | 4 | 2170 | ||
AF095193.2 | - | 4 | 2170 | |||
AF127139.1 | - | 4 | 2142 | |||
AK291333.1 | - | 4 | 1928 | |||
AL137582.1 | - | 4 | 1987 | |||
BC006418.1 | - | 4 | 2148 | 3635446 | ||
BC014656.1 | - | 4 | 1875 | 4636684 | ||
BC107786.1 | - | 4 | 1974 | 6655146 |