Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sfxn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022948.1 | NP_075237.1 | 3, 4 | 572 | ||
GenBank mRNA | AF276997.1 | - | 3, 4 | 572 | ||
BC062043.1 | - | 3, 4 | 601 | 5597953 |