Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022708.2 | NP_073199.1 | 1, 2 | 105 | ||
GenBank mRNA | AF054270.1 | - | 1, 2 | 105 |