Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P2ry12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022800.1 | NP_073637.1 | 2 | 659 | ||
XM_006232406.2 | XP_006232468.1 | 3 | 945 | |||
XM_006232407.2 | XP_006232469.1 | 3 | 1035 | |||
XM_006232408.2 | XP_006232470.1 | 3 | 2181 | |||
GenBank mRNA | AF313450.1 | - | 2 | 659 |
Set Membership: |
G-protein coupled receptor |