Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCB7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271696.1 | NP_001258625.1 | 16 | 2269 | ||
NM_001271697.1 | NP_001258626.1 | 15 | 2149 | |||
NM_001271698.1 | NP_001258627.1 | 15 | 2191 | |||
NM_001271699.1 | NP_001258628.1 | 15 | 2152 | |||
NM_004299.4 | NP_004290.2 | 16 | 2272 | |||
GenBank mRNA | AF038950.1 | - | 16 | 2209 | ||
AF078777.1 | - | 16 | 2189 | |||
AF133659.1 | - | 16 | 2201 | |||
BC006323.2 | - | 16 | 2205 | 4138236 | ||
BT009918.1 | - | 16 | 2204 | |||
BX537833.1 | - | 15 | 2089 | |||
CN411218.1 | - | 3 | 317 | |||
KU177872.1 | - | 16 | 2204 | |||
KU177873.1 | - | 14 | 1846 | |||
KU177874.1 | - | 6 | 770 | |||
KU177875.1 | - | 7 | 730 | |||
U66679.1 | - | 7 | 922 |