Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stx12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022939.2 | NP_075228.2 | 4 | 455 | ||
XM_006239066.2 | XP_006239128.1 | 4 | 499 | |||
GenBank mRNA | AF035632.1 | - | 4 | 371 | ||
CB581551.1 | - | 4 | 444 | |||
CK602227.1 | - | 4 | 455 |