Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022954.1 | NP_075243.1 | 2 | 427 | ||
GenBank mRNA | AB011527.1 | - | 2 | 427 |