Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024139.2 | NP_077053.1 | 3 | 301 | ||
GenBank mRNA | AB070350.1 | - | 3 | 286 | ||
BC062029.1 | - | 3 | 301 | 5623922 | ||
U39875.1 | - | 3 | 547 |