Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apex1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024148.1 | NP_077062.1 | 3 | 250 | ||
XM_006251913.1 | XP_006251975.1 | 3 | 291 | |||
GenBank mRNA | AF309114.1 | - | 2 | 166 | ||
AF311054.1 | - | 2 | 128 | |||
BC078816.1 | - | 3 | 258 | 7127205 | ||
D44495.1 | - | 3 | 250 | |||
FQ220521.1 | - | 3 | 252 | |||
L27076.1 | - | 3 | 234 |