Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DFFA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004401.2 | NP_004392.1 | 6 | 1286 | ||
NM_213566.1 | NP_998731.1 | 5 | 2641 | |||
GenBank mRNA | AK222546.1 | - | 6 | 1222 | ||
BC000037.2 | - | 6 | 1206 | 3505817 | ||
BC007112.1 | - | 6 | 1257 | 4292811 | ||
BC007721.2 | - | 6 | 1218 | 4299734 | ||
U91985.1 | - | 6 | 1244 |