Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270701.1 | NP_001257630.1 | 2, 3 | 399 | ||
NM_001270702.1 | NP_001257631.1 | 1, 3 | 312 | |||
NM_001270703.1 | NP_001257632.1 | 2, 3 | 399 | |||
NM_023021.2 | NP_075410.2 | 2, 3 | 399 | |||
XM_006228466.2 | XP_006228528.1 | 2, 3 | 2159 | |||
XM_006228467.2 | XP_006228529.1 | 2, 3 | 2160 | |||
XM_006228469.1 | XP_006228531.1 | 3, 4 | 381 | |||
XM_006228470.2 | XP_006228532.1 | 2, 3 | 229 | |||
XM_006228471.1 | XP_006228533.1 | 2, 3 | 2656 | |||
XM_008758975.1 | XP_008757197.1 | 3, 4 | 2054 | |||
XM_008758976.1 | XP_008757198.1 | 2, 3 | 578 | |||
XM_008758978.1 | XP_008757200.1 | 2, 3 | 218 | |||
GenBank mRNA | AB292803.1 | - | 2, 3 | 257 | ||
AF149250.1 | - | 2, 3 | 257 | |||
AF156554.1 | - | 2, 3 | 399 | |||
AF190458.1 | - | 2, 3 | 369 | |||
AJ133438.2 | - | 2, 3 | 257 | |||
BC128736.1 | - | 2, 3 | 350 | 8372655 | ||
EU872449.1 | - | 2, 3 | 372 | |||
EU872450.1 | - | 2, 3 | 372 | |||
EU872451.1 | - | 1, 3 | 285 |