Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DFFB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282669.1 | NP_001269598.1 | 7 | 1493 | ||
NM_001320132.1 | NP_001307061.1 | 7 | 1590 | |||
NM_001320136.1 | NP_001307065.1 | 7 | 1471 | |||
NM_004402.3 | NP_004393.1 | 7 | 1421 | |||
NR_104222.1 | 8 | 1558 | ||||
NR_135150.1 | 8 | 1545 | ||||
NR_135151.1 | 8 | 1508 | ||||
NR_135152.1 | 7 | 1443 | ||||
XM_017000498.1 | XP_016855987.1 | 6 | 1356 | |||
XM_017000499.1 | XP_016855988.1 | 6 | 1372 | |||
XM_017000500.1 | XP_016855989.1 | 6 | 1127 | |||
GenBank mRNA | AB028911.1 | - | 8 | 1338 | ||
AB028912.1 | - | 7 | 1384 | |||
AB028913.1 | - | 8 | 1302 | |||
AF064019.1 | - | 7 | 1215 | |||
AK290877.1 | - | 7 | 1212 | |||
AK303065.1 | - | 7 | 1300 | |||
BC032827.1 | - | 8 | 1370 | |||
BC048797.1 | - | 7 | 1407 | 5204996 |