Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myo1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023092.1 | NP_075580.2 | 6 | 812 | ||
GenBank mRNA | X74800.1 | - | 6 | 812 |