Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fgf20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023961.1 | NP_076451.1 | 3 | 427 | ||
GenBank mRNA | AB020021.1 | - | 3 | 427 |