Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DUSP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004417.3 | NP_004408.1 | 4 | 1417 | ||
GenBank mRNA | AK298047.1 | - | 5 | 1351 | ||
AK298658.1 | - | 5 | 1217 | |||
AK300487.1 | - | 5 | 1331 | |||
AK304475.1 | - | 5 | 1209 | |||
AK316321.1 | - | 5 | 1160 | |||
BC022463.1 | - | 4 | 1412 | 4794895 | ||
X68277.1 | - | 4 | 1402 |