Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp1r10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022951.2 | NP_075240.1 | 11 | 1468 | ||
XM_008772763.1 | XP_008770985.1 | 10 | 2152 | |||
XM_008772764.1 | XP_008770986.1 | 11 | 1950 | |||
GenBank mRNA | AF040954.1 | - | 10 | 1431 | ||
BC134805.1 | - | 10 | 1441 | 8362313 |