Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps33a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022961.3 | NP_075250.2 | 2, 3 | 192 | ||
GenBank mRNA | BC105752.1 | - | 2, 3 | 192 | 5599123 |