Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lphn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022962.1 | NP_075251.1 | 13 | 2462 | ||
GenBank mRNA | AF081144.1 | - | 12 | 2447 | ||
AF081145.1 | - | 12 | 2447 | |||
AF081146.1 | - | 13 | 2462 | |||
AF081147.1 | - | 13 | 2462 | |||
AF111099.1 | - | 14 | 2788 | |||
U72487.1 | - | 12 | 2264 | |||
U78105.1 | - | 12 | 2589 |
Set Membership: |
G-protein coupled receptor |