Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ESRRA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282450.1 | NP_001269379.1 | 7 | 1813 | ||
NM_001282451.1 | NP_001269380.1 | 7 | 1797 | |||
NM_004451.4 | NP_004442.3 | 7 | 1800 | |||
XM_017017313.1 | XP_016872802.1 | 6 | 2304 | |||
GenBank mRNA | BC007915.2 | - | 6 | 1270 | ||
BC011528.1 | - | 1 | 476 | |||
BC033701.1 | - | 5 | 1209 | |||
BC063795.1 | - | 7 | 1750 | 6527166 | ||
BC073956.1 | - | 3 | 666 | |||
BC092470.1 | - | 7 | 1813 | 30336569 | ||
L38487.1 | - | 7 | 1753 | |||
X51416.1 | - | 1937 |