Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lpar3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023969.1 | NP_076459.1 | 2 | 879 | ||
XM_006233469.2 | XP_006233531.1 | 2 | 919 | |||
XM_006233470.2 | XP_006233532.1 | 3 | 1087 | |||
XM_006233471.2 | XP_006233533.1 | 2 | 920 | |||
XM_008761522.1 | XP_008759744.1 | 3 | 1134 | |||
XM_008761523.1 | XP_008759745.1 | 3 | 1853 | |||
GenBank mRNA | AB051164.1 | - | 2 | 907 | ||
AF097733.1 | - | 2 | 879 |
Set Membership: |
G-protein coupled receptor |