Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc27a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024143.2 | NP_077057.1 | 8 | 1732 | ||
GenBank mRNA | AF242189.1 | - | 8 | 1733 | ||
BC091147.1 | - | 8 | 1755 | 7370589 |