Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024161.2 | NP_077075.1 | 3 | 407 | ||
XM_008762544.1 | XP_008760766.1 | 3 | 456 | |||
XM_008762545.1 | XP_008760767.1 | 4 | 497 | |||
GenBank mRNA | AF323955.1 | - | 3 | 407 | ||
FQ223796.1 | - | 3 | 448 | |||
S81917.1 | - | 2 | 307 | |||
U39320.1 | - | 3 | 407 |