Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mst1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024352.1 | NP_077328.1 | 2 | 124 | ||
XM_006243706.1 | XP_006243768.1 | 2 | 166 | |||
GenBank mRNA | BC089796.1 | - | 2 | 128 | 7322260 | |
X95096.1 | - | 2 | 124 |