Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nr4a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024388.2 | NP_077364.2 | 2 | 434 | ||
XM_006242356.2 | XP_006242418.1 | 2 | 3261 | |||
XM_006242358.2 | XP_006242420.1 | 2 | 1189 | |||
GenBank mRNA | BC097313.1 | - | 2 | 427 | 7458540 | |
CB581975.1 | - | 2 | 434 | |||
U17254.1 | - | 2 | 427 |