Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Htr3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024394.2 | NP_077370.2 | 3 | 366 | ||
GenBank mRNA | CB583270.1 | - | 3 | 366 | ||
D49395.1 | - | 3 | 257 |