Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnrpd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001082540.1 | NP_001076009.1 | 1, 2 | 509 | ||
NM_024404.2 | NP_077380.2 | 1, 2 | 509 | |||
GenBank mRNA | AB046615.1 | - | 1, 2 | 273 | ||
AB046617.1 | - | 1, 2 | 224 | |||
CO567918.1 | - | 1, 2 | 509 |