Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grpel1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024487.4 | NP_077813.1 | 2 | 213 | ||
XM_006251142.2 | XP_006251204.1 | 3 | 406 | |||
GenBank mRNA | BC097312.1 | - | 2 | 211 | 7387647 | |
FQ219742.1 | - | 3 | 394 | |||
FQ220295.1 | - | 2 | 204 | |||
U62940.1 | - | 2 | 214 |