Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IDI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317955.1 | NP_001304884.1 | 6 | 1597 | ||
NM_001317956.1 | NP_001304885.1 | 5 | 1731 | |||
NM_001317957.1 | NP_001304886.1 | 4 | 1558 | |||
NM_004508.3 | NP_004499.2 | 5 | 1614 | |||
NR_134300.1 | 4 | 1346 | ||||
NR_134301.1 | 4 | 1441 | ||||
XM_011519457.1 | XP_011517759.1 | 5 | 2461 | |||
XM_017016191.1 | XP_016871680.1 | 4 | 2288 | |||
GenBank mRNA | AF271720.1 | - | 4 | 1083 | ||
AK311110.1 | - | 4 | 992 | |||
BC005247.2 | - | 4 | 1114 | |||
BC006999.2 | - | 4 | 1114 | |||
BC019227.2 | - | 4 | 1114 | |||
BC022418.2 | - | 4 | 1114 | |||
BC025375.2 | - | 4 | 1113 | |||
BC057827.1 | - | 6 | 1587 | 30337530 | ||
BC107893.1 | - | 5 | 1140 | |||
BX537663.1 | - | 4 | 4160 | |||
BX648472.1 | - | 4 | 2077 | |||
X17025.1 | - | 5 | 1117 |