Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grk5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030829.1 | NP_110456.1 | 2 | 264 | ||
XM_006231686.1 | XP_006231748.1 | 2 | 317 | |||
GenBank mRNA | BC081792.1 | - | 2 | 317 | 7123515 | |
U34841.1 | - | 2 | 264 |