Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stxbp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030843.2 | NP_110470.1 | 4 | 753 | ||
NM_178345.1 | NP_848035.1 | 4 | 403 | |||
NM_178346.1 | NP_848036.1 | 4 | 403 | |||
XM_006227623.2 | XP_006227685.1 | 4 | 930 | |||
XM_006227624.2 | XP_006227686.1 | 4 | 930 | |||
XM_008758628.1 | XP_008756850.1 | 4 | 930 | |||
XM_008758629.1 | XP_008756851.1 | 4 | 930 | |||
GenBank mRNA | AF118889.1 | - | 4 | 403 | ||
AF118890.1 | - | 4 | 403 | |||
U92072.1 | - | 4 | 753 |