Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ILF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267809.1 | NP_001254738.1 | 10 | 817 | ||
NM_004515.3 | NP_004506.2 | 10 | 798 | |||
GenBank mRNA | AK223326.1 | - | 10 | 718 | ||
AK302214.1 | - | 10 | 761 | |||
AK313364.1 | - | 10 | 757 | |||
BC000382.2 | - | 10 | 723 | 2820505 | ||
BM800544.1 | - | 10 | 774 | |||
HQ322261.1 | - | 9 | 692 | |||
HQ322262.1 | - | 9 | 692 | |||
U10323.1 | - | 10 | 731 |