Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akap4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024402.1 | NP_077378.1 | 2, 3 | 173 | ||
GenBank mRNA | AF008114.1 | - | 2, 3 | 173 |