Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chrna1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024485.1 | NP_077811.1 | 7, 8 | 1071 | ||
GenBank mRNA | X74832.1 | - | 7, 8 | 1071 |