Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grpel1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024487.4 | NP_077813.1 | 2, 3 | 237 | ||
XM_006251142.2 | XP_006251204.1 | 3, 4 | 430 | |||
GenBank mRNA | BC097312.1 | - | 2, 3 | 235 | 7387647 | |
FQ219742.1 | - | 3, 4 | 418 | |||
FQ220295.1 | - | 2, 3 | 228 | |||
U62940.1 | - | 2, 3 | 238 |