Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slco1a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030838.1 | NP_110465.1 | 7 | 854 | ||
XM_006237642.2 | XP_006237704.1 | 9 | 1069 | |||
XM_006237643.2 | XP_006237705.1 | 9 | 1089 | |||
XM_006237646.2 | XP_006237708.1 | 7 | 870 | |||
XM_006237648.2 | XP_006237710.1 | 4 | 1282 | |||
XM_008763432.1 | XP_008761654.1 | 8 | 2438 | |||
GenBank mRNA | AF041105.1 | - | 6 | 617 | ||
AF083469.1 | - | 7 | 854 | |||
FQ211722.1 | - | 9 | 1084 |