Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fstl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024369.2 | NP_077345.1 | 11 | 996 | ||
GenBank mRNA | BC087014.1 | - | 11 | 996 | 7189359 | |
U06864.1 | - | 11 | 971 |