Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gabrg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024370.3 | NP_077346.3 | 6, 7 | 840 | ||
GenBank mRNA | M81142.1 | - | 6, 7 | 822 | ||
X63324.1 | - | 896 |