Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DAP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199849.1 | NP_001186778.1 | 7 | 589 | ||
NM_001199850.1 | NP_001186779.1 | 5 | 394 | |||
NM_001199851.1 | NP_001186780.1 | 5 | 373 | |||
NM_004632.3 | NP_004623.1 | 6 | 493 | |||
NM_033657.2 | NP_387506.1 | 6 | 562 | |||
XM_005245480.2 | XP_005245537.1 | 5 | 459 | |||
XM_005245481.2 | XP_005245538.1 | 5 | 438 | |||
XM_005245482.4 | XP_005245539.1 | 4 | 273 | |||
XM_011509972.2 | XP_011508274.1 | 6 | 492 | |||
XM_017002289.1 | XP_016857778.1 | 6 | 561 | |||
XM_017002290.1 | XP_016857779.1 | 7 | 585 | |||
XM_017002291.1 | XP_016857780.1 | 6 | 479 | |||
XM_017002292.1 | XP_016857781.1 | 6 | 458 | |||
XM_017002293.1 | XP_016857782.1 | 5 | 377 | |||
XM_017002294.1 | XP_016857783.1 | 6 | 479 | |||
XM_017002295.1 | XP_016857784.1 | 6 | 458 | |||
XM_017002296.1 | XP_016857785.1 | 5 | 438 | |||
XM_017002297.1 | XP_016857786.1 | 4 | 336 | |||
GenBank mRNA | AK225869.1 | - | 7 | 589 | ||
AK293117.1 | - | 6 | 494 | |||
AK298201.1 | - | 5 | 381 | |||
AK302281.1 | - | 5 | 394 | |||
BC002358.1 | - | 5 | 521 | |||
BC107487.1 | - | 6 | 493 | 40032516 | ||
BC107488.1 | - | 6 | 562 | 40032512 | ||
BT019494.1 | - | 5 | 405 | |||
CR749790.1 | - | 6 | 573 | |||
U18321.1 | - | 6 | 574 | |||
X83544.1 | - | 6 | 478 |