Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289978.1 | NP_001276907.1 | 11 | 1157 | ||
NM_004647.3 | NP_004638.2 | 10 | 1025 | |||
XM_006723407.2 | XP_006723470.1 | 11 | 1128 | |||
XM_006723408.3 | XP_006723471.1 | 11 | 1096 | |||
XM_006723409.2 | XP_006723472.1 | 11 | 1143 | |||
XM_006723410.2 | XP_006723473.1 | 8 | 695 | |||
XM_011527356.1 | XP_011525658.1 | 10 | 1106 | |||
XM_011527357.2 | XP_011525659.1 | 10 | 1046 | |||
XM_011527358.2 | XP_011525660.1 | 11 | 1172 | |||
GenBank mRNA | GQ358728.1 | - | 11 | 1049 | ||
U43843.1 | - | 10 | 923 |