Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Creb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031017.1 | NP_112279.1 | 5 | 383 | ||
NM_134443.1 | NP_604392.1 | 3 | 302 | |||
XM_006245065.2 | XP_006245127.1 | 5 | 389 | |||
XM_006245066.2 | XP_006245128.1 | 4 | 554 | |||
XM_006245067.2 | XP_006245129.1 | 4 | 294 | |||
XM_006245068.2 | XP_006245130.1 | 3 | 241 | |||
XM_006245069.2 | XP_006245131.1 | 3 | 891 | |||
XM_008767138.1 | XP_008765360.1 | 2 | 2745 | |||
GenBank mRNA | BC097311.1 | - | 5 | 583 | ||
FQ211925.1 | - | 5 | 448 | |||
X14788.1 | - | 5 | 383 | |||
X60002.1 | - | 3 | 302 |
Gene Symbol: LOC100363500 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008772050.1 | XP_008770272.1 | 1 | 337 |